4-Methyl-5-(5-methylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030295-500MG
101829-852EA 144.9 USD
101829-852
4-Methyl-5-(5-methylthien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Methyl-5-(5-methylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₉N₃S₂
MDL Number: MFCD03423415
CAS Number: 588685-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR