4,5-Diisopropyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030293-500MG
101829-848EA 193.85 USD
101829-848
4,5-Diisopropyl-4H-1,2,4-triazole-3-thiol
4,5-Diisopropyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₁₅N₃S
MDL Number: MFCD03423414
CAS Number: 667437-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR