5-Ethyl-4-isopropyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030287-500MG
101829-836EA 193.85 USD
101829-836
5-Ethyl-4-isopropyl-4H-1,2,4-triazole-3-thiol
5-Ethyl-4-isopropyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₇H₁₃N₃S
MDL Number: MFCD03423408
CAS Number: 869941-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR