5-Ethyl-4-propyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030286-500MG
101829-834EA 144.9 USD
101829-834
5-Ethyl-4-propyl-4H-1,2,4-triazole-3-thiol
5-Ethyl-4-propyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₇H₁₃N₃S
MDL Number: MFCD03423407
CAS Number: 869941-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR