4,5-Diethyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030284-500MG
101829-830EA 144.9 USD
101829-830
4,5-Diethyl-4H-1,2,4-triazole-3-thiol
4,5-Diethyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₆H₁₁N₃S
MDL Number: MFCD03423405
CAS Number: 29448-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR