5-Ethyl-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030283-500MG
101829-828EA 173.85 USD
101829-828
5-Ethyl-4-methyl-4H-1,2,4-triazole-3-thiol
5-Ethyl-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₅H₉N₃S
MDL Number: MFCD03423404
CAS Number: 4418-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR