5-Ethyl-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030282-500MG
101829-826EA 267.69 USD
101829-826
5-Ethyl-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol
5-Ethyl-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₅N₃OS
MDL Number: MFCD03423403
CAS Number: 869944-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR