4-Allyl-5-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030279-500MG
101829-820EA 144.9 USD
101829-820
4-Allyl-5-methyl-4H-1,2,4-triazole-3-thiol
4-Allyl-5-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₆H₉N₃S
MDL Number: MFCD02255623
CAS Number: 6232-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR