4,5-Dimethyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030277-500MG
101829-816EA 144.9 USD
101829-816
4,5-Dimethyl-4H-1,2,4-triazole-3-thiol
4,5-Dimethyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₄H₇N₃S
MDL Number: MFCD00598239
CAS Number: 38942-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR