4-Isothiocyanato-N-(tetrahydrofuran-2-ylmethyl)-benzenesulfonamide

Supplier: Matrix Scientific

030269-500MG
101829-804EA 144.9 USD
101829-804
4-Isothiocyanato-N-(tetrahydrofuran-2-ylmethyl)-benzenesulfonamide
4-Isothiocyanato-N-(tetrahydrofuran-2-ylmethyl)-benzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃S₂
MDL Number: MFCD09971971
CAS Number: 956576-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR