4-Isothiocyanato-N-(2-phenylethyl)-benzenesulfonamide

Supplier: Matrix Scientific

030257-500MG
101829-780EA 144.9 USD
101829-780
4-Isothiocyanato-N-(2-phenylethyl)-benzenesulfonamide
4-Isothiocyanato-N-(2-phenylethyl)-benzenesulfonamide

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₂S₂
MDL Number: MFCD09971959
CAS Number: 956576-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR