4-Isothiocyanato-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

030255-500MG
101829-776EA 336.92 USD
101829-776
4-Isothiocyanato-N-methylbenzenesulfonamide
4-Isothiocyanato-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₈H₈N₂O₂S₂
MDL Number: MFCD09971957
CAS Number: 223785-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR