N-(3,5-Dichlorophenyl)-4-isothiocyanatobenzenesulfonamide

Supplier: Matrix Scientific

030254-500MG
101829-774EA 144.9 USD
101829-774
N-(3,5-Dichlorophenyl)-4-isothiocyanatobenzenesulfonamide
N-(3,5-Dichlorophenyl)-4-isothiocyanatobenzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₈Cl₂N₂O₂S₂
MDL Number: MFCD09971956
CAS Number: 956576-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR