4-Isothiocyanato-N-(2-methylphenyl)benzenesulfonamide

Supplier: Matrix Scientific

030252-500MG
101829-770EA 193.85 USD
101829-770
4-Isothiocyanato-N-(2-methylphenyl)benzenesulfonamide
4-Isothiocyanato-N-(2-methylphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂S₂
MW: 304.39 g/mol
MDL Number: MFCD09971954
CAS Number: 107920-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR