2-[(4-Isothiocyanatophenyl)sulfonyl]-1,2,3,4-tetrahydroisoquinoline

Supplier: Matrix Scientific

030249-500MG
101829-764EA 144.9 USD
101829-764
2-[(4-Isothiocyanatophenyl)sulfonyl]-1,2,3,4-tetrahydroisoquinoline
2-[(4-Isothiocyanatophenyl)sulfonyl]-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₂S₂
MDL Number: MFCD09971952
CAS Number: 956576-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR