1-[(4-Isothiocyanatophenyl)sulfonyl]-3,5-dimethylpiperidine

Supplier: Matrix Scientific

030246-500MG
101829-758EA 144.9 USD
101829-758
1-[(4-Isothiocyanatophenyl)sulfonyl]-3,5-dimethylpiperidine
1-[(4-Isothiocyanatophenyl)sulfonyl]-3,5-dimethylpiperidine

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₂S₂
MDL Number: MFCD09971951
CAS Number: 956576-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR