4-Isothiocyanato-N,N-dimethylbenzenesulfonamide

Supplier: Matrix Scientific

030245-500MG
101829-756EA 307.69 USD
101829-756
4-Isothiocyanato-N,N-dimethylbenzenesulfonamide
4-Isothiocyanato-N,N-dimethylbenzenesulfonamide

Hazard: Irritant

Formula: C₉H₁₀N₂O₂S₂
MDL Number: MFCD09971950
CAS Number: 223785-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR