4-[(4-Isothiocyanatophenyl)sulfonyl]morpholine

Supplier: Matrix Scientific

030244-500MG
101829-754EA 144.9 USD
101829-754
4-[(4-Isothiocyanatophenyl)sulfonyl]morpholine
4-[(4-Isothiocyanatophenyl)sulfonyl]morpholine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S₂
MDL Number: MFCD03147328
CAS Number: 100060-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR