1-[(4-Isothiocyanatophenyl)sulfonyl]-2-methylpiperidine
Supplier: Matrix Scientific
030243-500MG
101829-752EA
193.85
USD
101829-752
1-[(4-Isothiocyanatophenyl)sulfonyl]-2-methylpiperidine
1-[(4-Isothiocyanatophenyl)sulfonyl]-2-methylpiperidine
Hazard: Irritant
Formula:
C₁₃H₁₆N₂O₂S₂ |
MDL Number:
MFCD09971949 CAS Number: 860440-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...