N-Ethyl-4-isothiocyanatobenzenesulfonamide

Supplier: Matrix Scientific

030242-500MG
101829-750EA 193.85 USD
101829-750
N-Ethyl-4-isothiocyanatobenzenesulfonamide
N-Ethyl-4-isothiocyanatobenzenesulfonamide

Hazard: Irritant

Formula: C₉H₁₀N₂O₂S₂
MDL Number: MFCD06194931
CAS Number: 726144-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR