1-[(4-Isothiocyanatophenyl)sulfonyl]-3-methylpiperidine

Supplier: Matrix Scientific

030240-500MG
101829-746EA 144.9 USD
101829-746
1-[(4-Isothiocyanatophenyl)sulfonyl]-3-methylpiperidine
1-[(4-Isothiocyanatophenyl)sulfonyl]-3-methylpiperidine

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂S₂
MDL Number: MFCD07626059
CAS Number: 111032-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR