1-[(4-Isothiocyanatophenyl)sulfonyl]-4-methylpiperidine

Supplier: Matrix Scientific

030238-500MG
101829-742EA 336.92 USD
101829-742
1-[(4-Isothiocyanatophenyl)sulfonyl]-4-methylpiperidine
1-[(4-Isothiocyanatophenyl)sulfonyl]-4-methylpiperidine

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂S₂
MDL Number: MFCD09971948
CAS Number: 111032-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR