1-[(4-Isothiocyanatophenyl)sulfonyl]azepane

Supplier: Matrix Scientific

030233-500MG
101829-732EA 193.85 USD
101829-732
1-[(4-Isothiocyanatophenyl)sulfonyl]azepane
1-[(4-Isothiocyanatophenyl)sulfonyl]azepane

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂S₂
MDL Number: MFCD09971943
CAS Number: 956576-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR