1-[(4-Isothiocyanatophenyl)sulfonyl]pyrrolidine

Supplier: Matrix Scientific

030232-500MG
101829-730EA 336.92 USD
101829-730
1-[(4-Isothiocyanatophenyl)sulfonyl]pyrrolidine
1-[(4-Isothiocyanatophenyl)sulfonyl]pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂S₂
MDL Number: MFCD09971942
CAS Number: 54286-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR