4-Isothiocyanato-N-(4-methoxyphenyl)benzenesulfonamide

Supplier: Matrix Scientific

030231-500MG
101829-728EA 193.85 USD
101829-728
4-Isothiocyanato-N-(4-methoxyphenyl)benzenesulfonamide
4-Isothiocyanato-N-(4-methoxyphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₃S₂
MW: 320.39 g/mol
MDL Number: MFCD09971941
CAS Number: 107922-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR