6-Isothiocyanato-2,3-dihydro-1,4-benzodioxine

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-1,4-benzodioxin-6-yl isothiocyanate

030227-500MG
101829-724EA 165.6 USD
101829-724
6-Isothiocyanato-2,3-dihydro-1,4-benzodioxine
6-Isothiocyanato-2,3-dihydro-1,4-benzodioxine

Hazard: Irritant

Formula: C₉H₇NO₂S
MW: 193.23 g/mol
MDL Number: MFCD00175465
CAS Number: 141492-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR