4-Isothiocyanato-N-1,3-thiazol-2-ylbenzenesulfonamide

Supplier: Matrix Scientific

030226-500MG
101829-722EA 336.92 USD
101829-722
4-Isothiocyanato-N-1,3-thiazol-2-ylbenzenesulfonamide
4-Isothiocyanato-N-1,3-thiazol-2-ylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₇N₃O₂S₃
MDL Number: MFCD03946160
CAS Number: 51908-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR