Ethyl-4-ethyl-2-isothiocyanato-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

030225-500MG
101829-720EA 217.35 USD
101829-720
Ethyl-4-ethyl-2-isothiocyanato-5-methylthiophene-3-carboxylate
Ethyl-4-ethyl-2-isothiocyanato-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₂S₂
MDL Number: MFCD03944637
CAS Number: 588695-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR