Dimethyl-5-isothiocyanato-3-methylthiophene-2,4-dicarboxylate

Supplier: Matrix Scientific

030223-500MG
101829-716EA 290.77 USD
101829-716
Dimethyl-5-isothiocyanato-3-methylthiophene-2,4-dicarboxylate
Dimethyl-5-isothiocyanato-3-methylthiophene-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₄S₂
MDL Number: MFCD09971939
CAS Number: 956576-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR