Methyl-5-[(dimethylamino)carbonyl]-2-isothiocyanato-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

030221-500MG
101829-712EA 217.35 USD
101829-712
Methyl-5-[(dimethylamino)carbonyl]-2-isothiocyanato-4-methylthiophene-3-carboxylate
Methyl-5-[(dimethylamino)carbonyl]-2-isothiocyanato-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S₂
MDL Number: MFCD03900493
CAS Number: 667436-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR