Ethyl-2-isothiocyanato-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

Supplier: Matrix Scientific

030218-500MG
101829-706EA 217.35 USD
101829-706
Ethyl-2-isothiocyanato-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Ethyl-2-isothiocyanato-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₅NO₂S₂
MDL Number: MFCD03945301
CAS Number: 257610-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR