Methyl-2-isothiocyanato-5-phenylthiophene-3-carboxylate

Supplier: Matrix Scientific

030213-500MG
101829-696EA 290.77 USD
101829-696
Methyl-2-isothiocyanato-5-phenylthiophene-3-carboxylate
Methyl-2-isothiocyanato-5-phenylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₉NO₂S₂
MDL Number: MFCD03945365
CAS Number: 588715-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR