Ethyl-2-isothiocyanato-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxylate

Supplier: Matrix Scientific

030212-500MG
101829-694EA 290.77 USD
101829-694
Ethyl-2-isothiocyanato-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxylate
Ethyl-2-isothiocyanato-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₇NO₂S₂
MDL Number: MFCD04058683
CAS Number: 669728-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR