Methyl-2-isothiocyanato-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

030209-500MG
101829-688EA 217.35 USD
101829-688
Methyl-2-isothiocyanato-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Methyl-2-isothiocyanato-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₂S₂
MDL Number: MFCD03945145
CAS Number: 588713-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR