Methyl-2-isothiocyanato-4,5-dimethylthiophene-3-carboxylate
Supplier: Matrix Scientific
030205-500MG
101829-680EA
290.77
USD
101829-680
Methyl-2-isothiocyanato-4,5-dimethylthiophene-3-carboxylate
Methyl-2-isothiocyanato-4,5-dimethylthiophene-3-carboxylate
Hazard: Irritant
Formula:
C₉H₉NO₂S₂ |
MDL Number:
MFCD03943783 CAS Number: 588676-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...