Methyl-5-ethyl-2-isothiocyanatothiophene-3-carboxylate

Supplier: Matrix Scientific

030204-500MG
101829-678EA 290.77 USD
101829-678
Methyl-5-ethyl-2-isothiocyanatothiophene-3-carboxylate
Methyl-5-ethyl-2-isothiocyanatothiophene-3-carboxylate

Hazard: Irritant

Formula: C₉H₉NO₂S₂
MDL Number: MFCD04969622
CAS Number: 869946-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR