Methyl-2-isothiocyanato-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

030203-500MG
101829-676EA 290.77 USD
101829-676
Methyl-2-isothiocyanato-5-methylthiophene-3-carboxylate
Methyl-2-isothiocyanato-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₈H₇NO₂S₂
MDL Number: MFCD03943713
CAS Number: 588675-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR