Methyl-2-isocyanato-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

030191-500MG
101829-662EA 332.35 USD
101829-662
Methyl-2-isocyanato-5-methylthiophene-3-carboxylate
Methyl-2-isocyanato-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₈H₇NO₃S
Boiling Pt: 105…110 °C (3 Torr)
Melting Pt: 42…45 °C
MDL Number: MFCD09971936
CAS Number: 956576-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR