2-Chloro-4-methylphenyl isocyanate

Supplier: Matrix Scientific

030190-500MG
101829-660EA 146.15 USD
101829-660
2-Chloro-4-methylphenyl isocyanate
2-Chloro-4-methylphenyl isocyanate

Hazard: Irritant

Formula: C₈H₆ClNO
CAS Number: 40398-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR