1-Chloro-5-isocyanato-2,4-dimethoxybenzene

Supplier: Matrix Scientific

030186-500MG
101829-652EA 146.15 USD
101829-652
1-Chloro-5-isocyanato-2,4-dimethoxybenzene
1-Chloro-5-isocyanato-2,4-dimethoxybenzene

Hazard: Irritant

Formula: C₉H₈ClNO₃
MDL Number: MFCD00013857
CAS Number: 55440-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR