6-Isocyanato-2,3-dihydro-1,4-benzodioxine

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-1,4-benzodioxin-6-yl isocyanate

030185-500MG
101829-650EA 269.23 USD
101829-650
6-Isocyanato-2,3-dihydro-1,4-benzodioxine
6-Isocyanato-2,3-dihydro-1,4-benzodioxine

Hazard: Irritant

Formula: C₉H₇NO₃
MW: 177.16 g/mol
MDL Number: MFCD02677705
CAS Number: 100275-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR