1-Isocyanato-2-methoxy-4-nitrobenzene
Supplier: Matrix Scientific
030177-500MG
101829-636EA
146.15
USD
101829-636
1-Isocyanato-2-methoxy-4-nitrobenzene
1-Isocyanato-2-methoxy-4-nitrobenzene
Hazard: Irritant
Formula:
C₈H₆N₂O₄ |
MDL Number:
MFCD00037066 CAS Number: 33484-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...