4-Chloro-2-isocyanato-1-methoxybenzene

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-methoxyphenyl isocyanate

030175-500MG
101829-632EA 60 USD
101829-632
4-Chloro-2-isocyanato-1-methoxybenzene
4-Chloro-2-isocyanato-1-methoxybenzene

Hazard: Toxic

Formula: C₈H₆ClNO₂
MW: 183.6 g/mol
MDL Number: MFCD00013860
CAS Number: 55440-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR