1-sec-Butyl-4-isocyanatobenzene

Supplier: Matrix Scientific

030171-500MG
101829-626EA 384.62 USD
101829-626
1-sec-Butyl-4-isocyanatobenzene
1-sec-Butyl-4-isocyanatobenzene

Hazard: Irritant

Formula: C₁₁H₁₃NO
Boiling Pt: 199…200 °C
Density: 0.988
MDL Number: MFCD03701601
CAS Number: 480439-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR