4-Ethylphenyl isocyanate

Supplier: Matrix Scientific

Synonyms: 1-Ethyl-4-isocyanatobenzene

030167-500MG
101829-618EA 109.25 USD
101829-618
4-Ethylphenyl isocyanate
4-Ethylphenyl isocyanate

Hazard: Irritant

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00013881
CAS Number: 23138-50-3
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR