Methyl-3-isocyanatobenzoate

Supplier: Matrix Scientific

Synonyms: 3-(Methoxycarbonyl)phenyl isocyanateMethyl3-isocyanatobenzoate

030163-500MG
101829-608EA 146.15 USD
101829-608
Methyl-3-isocyanatobenzoate
Methyl 3-isocyanatobenzoate

Hazard: Irritant

Formula: C₉H₇NO₃
MW: 177.16 g/mol
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR