4-Chloro-2-fluorophenyl isocyanate

Supplier: Matrix Scientific

030158-500MG
101829-594EA 109.25 USD
101829-594
4-Chloro-2-fluorophenyl isocyanate
4-Chloro-2-fluorophenyl isocyanate

Hazard: Irritant

Formula: C₇H₃ClFNO
CAS Number: 51163-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR