5-Isocyanatoindane

Supplier: Matrix Scientific

030156-500MG
101829-588EA 379.5 USD
101829-588
5-Isocyanatoindane
5-Isocyanatoindane

Hazard: Irritant

Formula: C₁₀H₉NO
MDL Number: MFCD01863687
CAS Number: 120912-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR