5-[(3-Acetylphenyl)amino]-5-oxopentanoic acid

Supplier: Matrix Scientific

030150-500MG
101829-578EA 146.15 USD
101829-578
5-[(3-Acetylphenyl)amino]-5-oxopentanoic acid
5-[(3-Acetylphenyl)amino]-5-oxopentanoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD00157633
CAS Number: 194784-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR