4-[(3-Acetylphenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

030149-500MG
101829-576EA 87.4 USD
101829-576
4-[(3-Acetylphenyl)amino]-4-oxobutanoic acid
4-[(3-Acetylphenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD01055653
CAS Number: 62134-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR